Learn More
Meso Scale Discovery R-PLEX Human Annexin A5 ASYQP
SDP
Catalog No. 5000079575
Encompass
The R-Plex Human Annexin A5 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures Annexin A5 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay allows for the quantitative measurement of Annexin A5 an important biomarker for apoptosis It can be used for research applications in oncology immunology cardiovascular disease and drug discovery and development The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrator
Catalog No. 50-000-79575 Supplier Meso Scale Discovery Supplier No. K151AGER22
The R-Plex Human Annexin A5 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures Annexin A5 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay allows for the quantitative measurement of Annexin A5 an important biomarker for apoptosis It can be used for research applications in oncology immunology cardiovascular disease and drug discovery and development The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrator