Learn More
Meso Scale Discovery R-PLEX Human ApoE ASY
SDP
Catalog No. 5000078333
Encompass
The Meso Scale Discovery R-PLEX Human APOE Assay is a multiplex immunoassay panel that measures Apolipoprotein E (APOE) in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides quantitative results in under 3 hours The panel includes a calibrator curve and controls to ensure accurate and reliable APOE quantification This assay is intended for research use only and requires the SECTOR Imager 2400 or SECTOR S 600 instrument for analysis With minimal sample volume requirements and a streamlined workflow this APOE panel enables high-throughput APOE profiling in biomarker research studies
Catalog No. 50-000-78333 Supplier Meso Scale Discovery Supplier No. K151AMLR2
Add to Cart
Add to Cart
The Meso Scale Discovery R-PLEX Human APOE Assay is a multiplex immunoassay panel that measures Apolipoprotein E (APOE) in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides quantitative results in under 3 hours The panel includes a calibrator curve and controls to ensure accurate and reliable APOE quantification This assay is intended for research use only and requires the SECTOR Imager 2400 or SECTOR S 600 instrument for analysis With minimal sample volume requirements and a streamlined workflow this APOE panel enables high-throughput APOE profiling in biomarker research studies