Learn More
Meso Scale Discovery R-PLEX Human ApoE ASYQP
SDP
Catalog No. 5000082439
Encompass
The Meso Scale Discovery R-PLEX Human APOE Assay is a multiplex immunoassay panel that measures Apolipoprotein E (APOE) in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides quantitative results for APOE with high sensitivity and a broad dynamic range The assay requires a small sample volume and provides results in as little as 3 5 hours The R-PLEX APOE Assay can be used to study APOE biology including lipid metabolism cardiovascular disease Alzheimer s disease and more This assay is for research use only
Catalog No. 50-000-82439 Supplier Meso Scale Discovery Supplier No. K151AMLR22
The Meso Scale Discovery R-PLEX Human APOE Assay is a multiplex immunoassay panel that measures Apolipoprotein E (APOE) in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides quantitative results for APOE with high sensitivity and a broad dynamic range The assay requires a small sample volume and provides results in as little as 3 5 hours The R-PLEX APOE Assay can be used to study APOE biology including lipid metabolism cardiovascular disease Alzheimer s disease and more This assay is for research use only