Learn More
Meso Scale Discovery R-PLEX Human AXL/UFO ASY
SDP
Catalog No. 5000079616
Encompass
The R-PLEX Human AXL UFO Assay from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of AXL in human serum plasma and cell culture supernatants It utilizes Meso Scale Diagnostics electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and lyophilized calibrator It can test up to 80 samples in duplicate and has a testing time of 3 5 hours
Catalog No. 50-000-79616 Supplier Meso Scale Discovery Supplier No. K151A5R2
The R-PLEX Human AXL UFO Assay from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of AXL in human serum plasma and cell culture supernatants It utilizes Meso Scale Diagnostics electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and lyophilized calibrator It can test up to 80 samples in duplicate and has a testing time of 3 5 hours