Learn More
Meso Scale Discovery R-PLEX Human AXL/UFO ASYQP
SDP
Catalog No. 5000079536
Encompass
The R-PLEX Human AXL UFO Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring AXL in human serum plasma and cell culture supernatants It uses electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The assay requires 50 L of sample and is read on the QuickPlex SQ 120 instrument The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags and solutions for a standard curve and performance assessment
Catalog No. 50-000-79536 Supplier Meso Scale Discovery Supplier No. K151A5R22
The R-PLEX Human AXL UFO Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring AXL in human serum plasma and cell culture supernatants It uses electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The assay requires 50 L of sample and is read on the QuickPlex SQ 120 instrument The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent tags and solutions for a standard curve and performance assessment