Learn More
Meso Scale Discovery R-PLEX Human B2M ASY
SDP
Catalog No. 5000081644
Encompass
The R-PLEX Human B2M Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring beta-2 microglobulin (B2M) in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision Requiring 50 L or less sample volume it performs up to 80 tests on two plates The kit includes a 96-well plate with capture antibody detection antibody buffers diluent and calibrators The assay time is 3 5 hours with a calibration range of 50-10 000 pg/mL
Catalog No. 50-000-81644 Supplier Meso Scale Discovery Supplier No. K151AKZR2
The R-PLEX Human B2M Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring beta-2 microglobulin (B2M) in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision Requiring 50 L or less sample volume it performs up to 80 tests on two plates The kit includes a 96-well plate with capture antibody detection antibody buffers diluent and calibrators The assay time is 3 5 hours with a calibration range of 50-10 000 pg/mL