Learn More
Meso Scale Discovery R-PLEX Human B2M ASYQP
SDP
Catalog No. 5000080591
Encompass
The R-PLEX Human B2M Assay is a multiplex immunoassay panel that measures Beta-2-Microglobulin in human serum plasma (EDTA heparin) and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology on the SECTOR Imager instrument The assay has a reportable range of 80-50 000 pg/mL and the minimum required sample volume is 25 L This product can aid in monitoring levels of B2M which may be elevated in conditions like chronic inflammation autoimmune disease and some cancers The R-PLEX assay provides high sensitivity requires low sample volume and allows for customization of biomarker panels
Catalog No. 50-000-80591 Supplier Meso Scale Discovery Supplier No. K151AKZR22
The R-PLEX Human B2M Assay is a multiplex immunoassay panel that measures Beta-2-Microglobulin in human serum plasma (EDTA heparin) and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology on the SECTOR Imager instrument The assay has a reportable range of 80-50 000 pg/mL and the minimum required sample volume is 25 L This product can aid in monitoring levels of B2M which may be elevated in conditions like chronic inflammation autoimmune disease and some cancers The R-PLEX assay provides high sensitivity requires low sample volume and allows for customization of biomarker panels