Learn More
Meso Scale Discovery R-PLEX Human B7-H6 ASY
SDP
Catalog No. 5000079622
Encompass
The R-PLEX Human B7-H6 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human B7-H6 and H6 in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate ECL-labeled detection antibodies calibrators and necessary reagents It provides reliable data with intra-assay CV 10% and inter-assay CV 15% and has a 9-month shelf life at 2-8 C This assay quantifies B7-H6 and H6 crucial in immune response
Catalog No. 50-000-79622 Supplier Meso Scale Discovery Supplier No. K151B1R2
The R-PLEX Human B7-H6 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human B7-H6 and H6 in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate ECL-labeled detection antibodies calibrators and necessary reagents It provides reliable data with intra-assay CV 10% and inter-assay CV 15% and has a 9-month shelf life at 2-8 C This assay quantifies B7-H6 and H6 crucial in immune response