Learn More
Meso Scale Discovery R-PLEX Human B7-H6 ASYQP
SDP
Catalog No. 5000081970
Encompass
The R-PLEX Human B7-H6 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human B7-H6 in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls diluents wash buffer read buffer and plate seals This assay helps quantify B7-H6 levels aiding in understanding immune function and disease progression
Catalog No. 50-000-81970 Supplier Meso Scale Discovery Supplier No. K151B1R22
The R-PLEX Human B7-H6 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human B7-H6 in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators controls diluents wash buffer read buffer and plate seals This assay helps quantify B7-H6 levels aiding in understanding immune function and disease progression