Learn More
Meso Scale Discovery R-PLEX Human Basigin ASY
SDP
Catalog No. 5000081972
Encompass
The R-PLEX Human Basigin Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring human Basigin (CD147/EMMPRIN) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection for high sensitivity 5-logs dynamic range and low sample volume The kit includes a 96-well plate with capture antibody detection antibody calibrator and diluent It detects Basigin from 0 05-500 ng/mL with 25 L sample per well enabling reliable quantification of Basigin crucial in reproductive biology and tumor metastasis
Catalog No. 50-000-81972 Supplier Meso Scale Discovery Supplier No. K151ALVR2
The R-PLEX Human Basigin Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring human Basigin (CD147/EMMPRIN) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection for high sensitivity 5-logs dynamic range and low sample volume The kit includes a 96-well plate with capture antibody detection antibody calibrator and diluent It detects Basigin from 0 05-500 ng/mL with 25 L sample per well enabling reliable quantification of Basigin crucial in reproductive biology and tumor metastasis