Learn More
Meso Scale Discovery R-PLEX Human Basigin ASYQP
SDP
Catalog No. 5000080161
Encompass
The Meso Scale Discovery Basigin Kit is an electrochemiluminescent immunoassay designed for the quantitative measurement of Basigin in human serum plasma and cell culture supernatants The assay utilizes MSD s patented SULFO-TAG labels which emit light when electrochemically stimulated via carbon electrodes in the bottom of each well The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrator and sample diluent The assay has a typical sensitivity of 0 03 ng/mL and an upper limit of quantification of 50 ng/mL The total assay time is approximately 3 5 hours
Catalog No. 50-000-80161 Supplier Meso Scale Discovery Supplier No. K151ALVR22
The Meso Scale Discovery Basigin Kit is an electrochemiluminescent immunoassay designed for the quantitative measurement of Basigin in human serum plasma and cell culture supernatants The assay utilizes MSD s patented SULFO-TAG labels which emit light when electrochemically stimulated via carbon electrodes in the bottom of each well The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrator and sample diluent The assay has a typical sensitivity of 0 03 ng/mL and an upper limit of quantification of 50 ng/mL The total assay time is approximately 3 5 hours