Learn More
Meso Scale Discovery R-PLEX Human BCA-1/BLC ASYQP
SDP
Catalog No. 5000079460
Encompass
The R-PLEX Human BCA-1 (BLC) Assay from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of B-Cell Activating Factor (BAFF also known as B Lymphocyte Stimulator (BLyS)) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides a sensitive and reproducible method for measuring BAFF levels with a detection range of 1 95 - 1000 pg/mL The assay requires 50 L of sample and can be completed in under 5 hours This product could be useful for research related to autoimmune diseases immune monitoring or biomarker discovery and validation
Catalog No. 50-000-79460 Supplier Meso Scale Discovery Supplier No. K1519QR22
The R-PLEX Human BCA-1 (BLC) Assay from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of B-Cell Activating Factor (BAFF also known as B Lymphocyte Stimulator (BLyS)) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides a sensitive and reproducible method for measuring BAFF levels with a detection range of 1 95 - 1000 pg/mL The assay requires 50 L of sample and can be completed in under 5 hours This product could be useful for research related to autoimmune diseases immune monitoring or biomarker discovery and validation