Learn More
Meso Scale Discovery R-PLEX Human BChE ASY
SDP
Catalog No. 5000078869
Encompass
The R-PLEX Human BCHE Assay Kit from Meso Scale Diagnostics measures butyrylcholinesterase (BCHE) in human samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive BCHE detection a biomarker for organophosphorus pesticide and nerve agent exposure The kit includes a 96-well plate detection antibody with ruthenium label calibrators and reagents for 50 wells Results are analyzed with MSD s systems offering high sensitivity and minimal sample volume
Catalog No. 50-000-78869 Supplier Meso Scale Discovery Supplier No. K151AJCR2
The R-PLEX Human BCHE Assay Kit from Meso Scale Diagnostics measures butyrylcholinesterase (BCHE) in human samples This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive BCHE detection a biomarker for organophosphorus pesticide and nerve agent exposure The kit includes a 96-well plate detection antibody with ruthenium label calibrators and reagents for 50 wells Results are analyzed with MSD s systems offering high sensitivity and minimal sample volume