Learn More
Meso Scale Discovery R-PLEX Human BChE ASYQP
SDP
Catalog No. 5000081661
Encompass
The R-PLEX Human BCHE Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures butyrylcholinesterase (BCHE) in human serum plasma and other biological fluids This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 8-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals sufficient for one 96-well plate This assay can be used to measure BCHE levels and activity which may be altered in certain disease states
Catalog No. 50-000-81661 Supplier Meso Scale Discovery Supplier No. K151AJCR22
The R-PLEX Human BCHE Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures butyrylcholinesterase (BCHE) in human serum plasma and other biological fluids This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 8-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals sufficient for one 96-well plate This assay can be used to measure BCHE levels and activity which may be altered in certain disease states