Learn More
Meso Scale Discovery R-PLEX Human Bcl-2 ASY
SDP
Catalog No. 5000080315
Encompass
The R-PLEX Human Bcl-2 Assay Kit from Meso Scale Discovery is used to quantitatively measure Bcl-2 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection combined with patterned arrays The assay has high sensitivity a broad dynamic range and requires low sample volume It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker A calibration standard and control lysates The kit can test up to 80 samples in duplicate and has a testing time of 3 5 hours
Catalog No. 50-000-80315 Supplier Meso Scale Discovery Supplier No. K1516YR2
The R-PLEX Human Bcl-2 Assay Kit from Meso Scale Discovery is used to quantitatively measure Bcl-2 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection combined with patterned arrays The assay has high sensitivity a broad dynamic range and requires low sample volume It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker A calibration standard and control lysates The kit can test up to 80 samples in duplicate and has a testing time of 3 5 hours