Learn More
Meso Scale Discovery R-PLEX Human Bcl-2 ASYQP
SDP
Catalog No. 5000078886
Encompass
The R-PLEX Human Bcl-2 Assay Kit from Meso Scale Discovery is used to quantitatively measure Bcl-2 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection combined with patterned arrays The assay has high sensitivity requires low sample volume and has a fast turnaround time It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker A calibration standard and control lysates The kit can test up to 80 samples in duplicates and has a testing time of 3 5 hours
Catalog No. 50-000-78886 Supplier Meso Scale Discovery Supplier No. K1516YR22
The R-PLEX Human Bcl-2 Assay Kit from Meso Scale Discovery is used to quantitatively measure Bcl-2 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection combined with patterned arrays The assay has high sensitivity requires low sample volume and has a fast turnaround time It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker A calibration standard and control lysates The kit can test up to 80 samples in duplicates and has a testing time of 3 5 hours