Learn More
Meso Scale Discovery R-PLEX Human Bikunin ASY
SDP
Catalog No. 5000081625
Encompass
The R-PLEX Human Bikunin Assay from Meso Scale Diagnostics measures human bikunin in samples like serum plasma urine and cell lysates It uses electrochemiluminescence for high sensitivity and a broad range needing low sample volume The assay requiring an MSD instrument takes about 4 hours and detects bikunin from 0 05 - 1000 ng/mL Bikunin is a serine protease inhibitor in inflammatory cascades The kit includes a 96-well plate with capture antibody detection antibody calibrator diluent wash buffer read buffer and a plate seal
Catalog No. 50-000-81625 Supplier Meso Scale Discovery Supplier No. K151ALBR2
The R-PLEX Human Bikunin Assay from Meso Scale Diagnostics measures human bikunin in samples like serum plasma urine and cell lysates It uses electrochemiluminescence for high sensitivity and a broad range needing low sample volume The assay requiring an MSD instrument takes about 4 hours and detects bikunin from 0 05 - 1000 ng/mL Bikunin is a serine protease inhibitor in inflammatory cascades The kit includes a 96-well plate with capture antibody detection antibody calibrator diluent wash buffer read buffer and a plate seal