Learn More
Meso Scale Discovery R-PLEX Human BIM/Bcl2-L-11 ASY
SDP
Catalog No. 5000080254
Encompass
The R-PLEX Human BIM/BCL2-L-11 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures the levels of BIM and BCL2-L-11 proteins in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides sensitive accurate and reproducible quantitation of these apoptosis-related proteins The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and controls Results are obtained in as little as 3 5 hours from sample addition This assay allows for the simultaneous measurement of multiple biomarkers related to apoptosis regulation
Catalog No. 50-000-80254 Supplier Meso Scale Discovery Supplier No. K1517AR2
The R-PLEX Human BIM/BCL2-L-11 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures the levels of BIM and BCL2-L-11 proteins in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides sensitive accurate and reproducible quantitation of these apoptosis-related proteins The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and controls Results are obtained in as little as 3 5 hours from sample addition This assay allows for the simultaneous measurement of multiple biomarkers related to apoptosis regulation