Learn More
Meso Scale Discovery R-PLEX Human BMP-2 ASY
SDP
Catalog No. 5000079578
Encompass
The R-PLEX Human BMP-2 Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human Bone Morphogenetic Protein 2 (BMP-2) in cell culture supernatants serum plasma and other biological matrices The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and rapid detection of BMP-2 with a lower limit of detection of 1 5 pg/mL and an assay range of up to 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrators and all necessary reagents for the quantitative measurement of BMP-2 in approximately 4 hours
Catalog No. 50-000-79578 Supplier Meso Scale Discovery Supplier No. K1519RR2
The R-PLEX Human BMP-2 Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human Bone Morphogenetic Protein 2 (BMP-2) in cell culture supernatants serum plasma and other biological matrices The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and rapid detection of BMP-2 with a lower limit of detection of 1 5 pg/mL and an assay range of up to 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrators and all necessary reagents for the quantitative measurement of BMP-2 in approximately 4 hours