Learn More
Meso Scale Discovery R-PLEX Human BMP-4 ASY
SDP
Catalog No. 5000079448
Encompass
The R-PLEX Human BMP-4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the concentration of human Bone Morphogenetic Protein 4 (BMP-4) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and control The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79448 Supplier Meso Scale Discovery Supplier No. K1519SR2
Add to Cart
Add to Cart
The R-PLEX Human BMP-4 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the concentration of human Bone Morphogenetic Protein 4 (BMP-4) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and control The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL