Learn More
Meso Scale Discovery R-PLEX Human BMP-5 ASY
SDP
Catalog No. 5000078963
Encompass
The R-PLEX Human BMP-5 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures BMP-5 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and lyophilized BMP-5 calibrator The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It can be used to measure BMP-5 levels in biological samples for research applications
Catalog No. 50-000-78963 Supplier Meso Scale Discovery Supplier No. K151AAFR2
The R-PLEX Human BMP-5 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures BMP-5 levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and lyophilized BMP-5 calibrator The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It can be used to measure BMP-5 levels in biological samples for research applications