Learn More
Meso Scale Discovery R-PLEX Human BMP-6 ASY
SDP
Catalog No. 5000079620
Encompass
The R-PLEX Human BMP-6 Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human Bone Morphogenetic Protein 6 (BMP-6) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager 6000 for high sensitivity detection The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent The assay has a range from 1 5 pg/mL to 1000 pg/mL and takes approximately 3 5 hours to complete This product can be used for biomarker research related to bone formation fracture healing and developmental disorders
Catalog No. 50-000-79620 Supplier Meso Scale Discovery Supplier No. K151AAGR2
The R-PLEX Human BMP-6 Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of human Bone Morphogenetic Protein 6 (BMP-6) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager 6000 for high sensitivity detection The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent The assay has a range from 1 5 pg/mL to 1000 pg/mL and takes approximately 3 5 hours to complete This product can be used for biomarker research related to bone formation fracture healing and developmental disorders