Learn More
Meso Scale Discovery R-PLEX Human BMP-7 ASY
SDP
Catalog No. 5000078501
Encompass
The R-PLEX Human BMP-7 Assay Kit from Meso Scale Discovery is used to quantitatively measure BMP-7 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection combined with patterned arrays The assay has high sensitivity and a broad dynamic range It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and a vial of calibrator The kit has a measuring range of 1 95 - 2500 pg/mL and requires 25 L of sample
Catalog No. 50-000-78501 Supplier Meso Scale Discovery Supplier No. K151D1R2
The R-PLEX Human BMP-7 Assay Kit from Meso Scale Discovery is used to quantitatively measure BMP-7 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection combined with patterned arrays The assay has high sensitivity and a broad dynamic range It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator diluent and a vial of calibrator The kit has a measuring range of 1 95 - 2500 pg/mL and requires 25 L of sample