Learn More
Meso Scale Discovery R-PLEX Human BTLA ASY
SDP
Catalog No. 5000081946
Encompass
The R-PLEX Human BTLA Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that enables simultaneous quantitative measurement of BTLA in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the precise and reproducible quantification of BTLA which plays an important role in regulating T cell activation With a rapid 3 5 hour protocol minimal hands-on time is required
Catalog No. 50-000-81946 Supplier Meso Scale Discovery Supplier No. K151ALNR2
The R-PLEX Human BTLA Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that enables simultaneous quantitative measurement of BTLA in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the precise and reproducible quantification of BTLA which plays an important role in regulating T cell activation With a rapid 3 5 hour protocol minimal hands-on time is required