Learn More
Meso Scale Discovery R-PLEX Human C1q receptor ASY
SDP
Catalog No. 5000078964
Encompass
The Meso Scale Discovery R-PLEX Human C1q Receptor Assay is a multiplex immunoassay panel that measures C1q Receptor in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay is available as a singleplex or as a multiplex combining up to 10 biomarkers per well It requires 50 L of sample and can perform up to 80 tests across two plates The assay takes 3 5 hours to complete and has a typical sensitivity of 19 pg/mL
Catalog No. 50-000-78964 Supplier Meso Scale Discovery Supplier No. K151AAHR2
The Meso Scale Discovery R-PLEX Human C1q Receptor Assay is a multiplex immunoassay panel that measures C1q Receptor in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay is available as a singleplex or as a multiplex combining up to 10 biomarkers per well It requires 50 L of sample and can perform up to 80 tests across two plates The assay takes 3 5 hours to complete and has a typical sensitivity of 19 pg/mL