Learn More
Meso Scale Discovery R-PLEX Human CA125 ASY
SDP
Catalog No. 5000078931
Encompass
The Meso Scale Discovery R-PLEX Human CA125 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of CA125 in human serum plasma and cell culture supernatant It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The assay has a range of 0 6 - 2500 U/mL and includes reagents for one 96-well plate This product can aid in monitoring disease progression and response to therapy for ovarian cancer The kit includes a plate detection antibody sample diluent calibrator diluent wash buffer read buffer and a vial of SULFO-TAG labeled detection antibody
Catalog No. 50-000-78931 Supplier Meso Scale Discovery Supplier No. K151YVR2
The Meso Scale Discovery R-PLEX Human CA125 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of CA125 in human serum plasma and cell culture supernatant It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The assay has a range of 0 6 - 2500 U/mL and includes reagents for one 96-well plate This product can aid in monitoring disease progression and response to therapy for ovarian cancer The kit includes a plate detection antibody sample diluent calibrator diluent wash buffer read buffer and a vial of SULFO-TAG labeled detection antibody