Learn More
Meso Scale Discovery R-PLEX Human CA125 ASYQP
SDP
Catalog No. 5000078787
Encompass
The R-PLEX Human CA125 Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for measuring CA125 antigen in human serum plasma and cell culture supernatant It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results With a sensitivity of 0 2 U/mL and a range of 0 2-1500 U/mL it quantifies CA125 levels An MSD instrument like QuickPlex is required to read the plate The assay offers a fast simple and robust method for measuring CA125 in serum and plasma
Catalog No. 50-000-78787 Supplier Meso Scale Discovery Supplier No. K151YVR22
The R-PLEX Human CA125 Assay from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for measuring CA125 antigen in human serum plasma and cell culture supernatant It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results With a sensitivity of 0 2 U/mL and a range of 0 2-1500 U/mL it quantifies CA125 levels An MSD instrument like QuickPlex is required to read the plate The assay offers a fast simple and robust method for measuring CA125 in serum and plasma