Learn More
Meso Scale Discovery R-PLEX Human Caspase-1 ASY
SDP
Catalog No. 5000082347
Encompass
The R-Plex Human Caspase-1 Assay Kit from Meso Scale Discovery is used to quantitatively measure caspase-1 in human biological samples such as serum plasma cell culture supernatants cell lysates tissue homogenates or extracts The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate coated with capture antibody detection antibody labeled with an ECL tag assay diluent calibrator diluent wash buffer read buffer and a vial of calibrator The total assay time is 3 5 hours for 96 wells The kit can detect caspase-1 concentrations as low as 17 pg/mL
Catalog No. 50-000-82347 Supplier Meso Scale Discovery Supplier No. K151K8R2
The R-Plex Human Caspase-1 Assay Kit from Meso Scale Discovery is used to quantitatively measure caspase-1 in human biological samples such as serum plasma cell culture supernatants cell lysates tissue homogenates or extracts The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate coated with capture antibody detection antibody labeled with an ECL tag assay diluent calibrator diluent wash buffer read buffer and a vial of calibrator The total assay time is 3 5 hours for 96 wells The kit can detect caspase-1 concentrations as low as 17 pg/mL