Learn More
Meso Scale Discovery R-PLEX Human Caspase-1 ASYQP
SDP
Catalog No. 5000080798
Encompass
The R-Plex Human Caspase-1 Assay Kit from Meso Scale Discovery is used to quantitatively measure caspase-1 in human biological samples such as serum plasma cell culture supernatants and cell lysates The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag and calibrator diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation period The kit can perform up to 94 assays in the 96-well plate format
Catalog No. 50-000-80798 Supplier Meso Scale Discovery Supplier No. K151K8R22
The R-Plex Human Caspase-1 Assay Kit from Meso Scale Discovery is used to quantitatively measure caspase-1 in human biological samples such as serum plasma cell culture supernatants and cell lysates The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag and calibrator diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation period The kit can perform up to 94 assays in the 96-well plate format