Learn More
Meso Scale Discovery R-PLEX Human Cathepsin D ASYQP
SDP
Catalog No. 5000081844
Encompass
The R-PLEX Human Cathepsin D Assay Kit from Meso Scale Discovery is used to quantitatively measure levels of active human Cathepsin D in cell culture supernatants serum plasma or other biological samples This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and rapid detection of Cathepsin D with a lower limit of detection of 19 pg/mL The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium label calibrators and controls The assay can be fully automated on MSD instrument platforms and has a total assay time of 3 5 hours
Catalog No. 50-000-81844 Supplier Meso Scale Discovery Supplier No. K151ZZR22
The R-PLEX Human Cathepsin D Assay Kit from Meso Scale Discovery is used to quantitatively measure levels of active human Cathepsin D in cell culture supernatants serum plasma or other biological samples This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and rapid detection of Cathepsin D with a lower limit of detection of 19 pg/mL The kit includes a 96-well plate pre-coated with capture antibody detection antibody conjugated to ruthenium label calibrators and controls The assay can be fully automated on MSD instrument platforms and has a total assay time of 3 5 hours