Learn More
Meso Scale Discovery R-PLEX Human Cathepsin L2 ASY
SDP
Catalog No. 5000078867
Encompass
The R-Plex Human Cathepsin L2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Cathepsin L2 protein in cell and tissue lysates It uses a quantitative sandwich immunoassay technique and color-coded magnetic beads for multiplex analysis The assay has a detection range of 78-50 000 pg/mL and includes a 96-well plate assay buffer wash buffer serum matrix standards detection antibodies streptavidin-PE and magnetic beads It can help researchers study Cathepsin L2 s role in cancer bone remodeling and protein processing The assay requires a Luminex-based reader and xPONENT software for analysis
Catalog No. 50-000-78867 Supplier Meso Scale Discovery Supplier No. K151AJGR2
Add to Cart
Add to Cart
The R-Plex Human Cathepsin L2 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Cathepsin L2 protein in cell and tissue lysates It uses a quantitative sandwich immunoassay technique and color-coded magnetic beads for multiplex analysis The assay has a detection range of 78-50 000 pg/mL and includes a 96-well plate assay buffer wash buffer serum matrix standards detection antibodies streptavidin-PE and magnetic beads It can help researchers study Cathepsin L2 s role in cancer bone remodeling and protein processing The assay requires a Luminex-based reader and xPONENT software for analysis