Learn More
Meso Scale Discovery R-PLEX Human CCN1 ASYQP
SDP
Catalog No. 5000080162
Encompass
The R-PLEX Human CCN1 Assay is a multiplex immunoassay kit for measuring CCN1 protein in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) on the Meso Scale Discovery (MSD) platform for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent and wash buffer Sample incubation is 2 hours with results in 3 5 hours Designed for research use only it requires an MSD instrument The kit offers high sensitivity accuracy and rapid results for quantifying CCN1 levels
Catalog No. 50-000-80162 Supplier Meso Scale Discovery Supplier No. K151ALYR22
The R-PLEX Human CCN1 Assay is a multiplex immunoassay kit for measuring CCN1 protein in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) on the Meso Scale Discovery (MSD) platform for sensitive detection The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent and wash buffer Sample incubation is 2 hours with results in 3 5 hours Designed for research use only it requires an MSD instrument The kit offers high sensitivity accuracy and rapid results for quantifying CCN1 levels