Learn More
Meso Scale Discovery R-PLEX Human CD14 ASY
SDP
Catalog No. 5000082300
Encompass
The R-PLEX Human CD14 Assay Kit from Meso Scale Discovery measures CD14 in human serum plasma and cell culture supernatants using electrochemiluminescence detection It offers high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrators and controls for a standard curve This quantitative assay takes about 4 hours and is ideal for immunology and inflammation research
Catalog No. 50-000-82300 Supplier Meso Scale Discovery Supplier No. K151T1R2
The R-PLEX Human CD14 Assay Kit from Meso Scale Discovery measures CD14 in human serum plasma and cell culture supernatants using electrochemiluminescence detection It offers high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrators and controls for a standard curve This quantitative assay takes about 4 hours and is ideal for immunology and inflammation research