Learn More
Meso Scale Discovery R-PLEX Human CD44 ASYQP
SDP
Catalog No. 5000080152
Encompass
The R-PLEX Human CD44 Assay is a multiplex immunoassay kit used to measure CD44 protein levels in human samples such as serum plasma cell culture supernatants cell lysates or other biological fluids The assay utilizes microsphere beads conjugated with anti-CD44 antibodies to capture CD44 proteins which are then detected using a biotinylated anti-CD44 antibody and streptavidin-phycoerythrin The kit includes reagents and buffers needed to run the assay and analyze CD44 levels using the Luminex detection platform The R-PLEX assay allows for the simultaneous detection of multiple proteins in a single small sample volume
Catalog No. 50-000-80152 Supplier Meso Scale Discovery Supplier No. K151AJHR22
The R-PLEX Human CD44 Assay is a multiplex immunoassay kit used to measure CD44 protein levels in human samples such as serum plasma cell culture supernatants cell lysates or other biological fluids The assay utilizes microsphere beads conjugated with anti-CD44 antibodies to capture CD44 proteins which are then detected using a biotinylated anti-CD44 antibody and streptavidin-phycoerythrin The kit includes reagents and buffers needed to run the assay and analyze CD44 levels using the Luminex detection platform The R-PLEX assay allows for the simultaneous detection of multiple proteins in a single small sample volume