Learn More
Meso Scale Discovery R-PLEX Human CD47 ASY
SDP
Catalog No. 5000082432
Encompass
The R-PLEX Human CD47 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring CD47 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for simultaneous detection with other biomarkers The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents The detection limit is 0 03 ng/mL with a range of 0 03-400 ng/mL enabling high-throughput sensitive and reproducible quantification of soluble CD47
Catalog No. 50-000-82432 Supplier Meso Scale Discovery Supplier No. K151ALZR2
The R-PLEX Human CD47 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring CD47 in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for simultaneous detection with other biomarkers The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents The detection limit is 0 03 ng/mL with a range of 0 03-400 ng/mL enabling high-throughput sensitive and reproducible quantification of soluble CD47