Learn More
Meso Scale Discovery R-PLEX Human CD5 ASY
SDP
Catalog No. 5000080945
Encompass
The R-PLEX Human CD5 Assay from Meso Scale Diagnostics is a multiplex immunoassay for measuring CD5 in human serum plasma (EDTA heparin) and cell culture supernatants It uses electrochemiluminescence (ECL) detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs It quantifies CD5 from 0 61 pg/mL to 10 000 pg/mL with high specificity and minimal cross-reactivity The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls enabling efficient and accurate CD5 quantification important for T cell activation and differentiation
Catalog No. 50-000-80945 Supplier Meso Scale Discovery Supplier No. K1563JR2
The R-PLEX Human CD5 Assay from Meso Scale Diagnostics is a multiplex immunoassay for measuring CD5 in human serum plasma (EDTA heparin) and cell culture supernatants It uses electrochemiluminescence (ECL) detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs It quantifies CD5 from 0 61 pg/mL to 10 000 pg/mL with high specificity and minimal cross-reactivity The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls enabling efficient and accurate CD5 quantification important for T cell activation and differentiation