Learn More
Meso Scale Discovery R-PLEX Human CD5 ASYQP
SDP
Catalog No. 5000081025
Encompass
The R-PLEX Human CD5 Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring CD5 in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators and necessary reagents It aids in monitoring CD5 levels and T cell activation allowing rapid and accurate quantification with minimal hands-on time
Catalog No. 50-000-81025 Supplier Meso Scale Discovery Supplier No. K1563JR22
The R-PLEX Human CD5 Assay from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring CD5 in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies calibrators and necessary reagents It aids in monitoring CD5 levels and T cell activation allowing rapid and accurate quantification with minimal hands-on time