Learn More
Meso Scale Discovery R-PLEX Human CD5L ASY
SDP
Catalog No. 5000080543
Encompass
The R-PLEX Human CD5L Assay from Meso Scale Diagnostics is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of CD5L in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes a CD5L-specific antibody coated plate detection antibody calibrator and assay diluent The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of quantification of 2500 ng/mL for CD5L This product can aid in the quantification of CD5L for life science research applications
Catalog No. 50-000-80543 Supplier Meso Scale Discovery Supplier No. K151T0R2
The R-PLEX Human CD5L Assay from Meso Scale Diagnostics is a multiplex immunoassay kit that enables the simultaneous quantitative measurement of CD5L in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes a CD5L-specific antibody coated plate detection antibody calibrator and assay diluent The assay has a typical sensitivity of 0 3 ng/mL and an upper limit of quantification of 2500 ng/mL for CD5L This product can aid in the quantification of CD5L for life science research applications