Learn More
Meso Scale Discovery R-PLEX Human CD80/B7-1 ASY
SDP
Catalog No. 5000078911
Encompass
The R-PLEX Human CD80 (B7-1) Assay Kit from Meso Scale Discovery measures CD80 levels in human serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection It offers high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture and detection antibodies buffers blocker solutions calibrator and controls Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL suitable for biomarker discovery and immunology research
Catalog No. 50-000-78911 Supplier Meso Scale Discovery Supplier No. K1518WR2
The R-PLEX Human CD80 (B7-1) Assay Kit from Meso Scale Discovery measures CD80 levels in human serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection It offers high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture and detection antibodies buffers blocker solutions calibrator and controls Sensitivity is 0 3 pg/mL with an upper quantification limit of 10 000 pg/mL suitable for biomarker discovery and immunology research