Learn More
Meso Scale Discovery R-PLEX Human CD80/B7-1 ASYQP
SDP
Catalog No. 5000080316
Encompass
The R-PLEX Human CD80 (B7-1) Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the human CD80 protein in serum plasma cell culture supernatants cell and tissue lysates and other biological samples The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80316 Supplier Meso Scale Discovery Supplier No. K1518WR22
The R-PLEX Human CD80 (B7-1) Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures the human CD80 protein in serum plasma cell culture supernatants cell and tissue lysates and other biological samples The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL