Learn More
Meso Scale Discovery R-PLEX Human CD81 EV ASY
SDP
Catalog No. 5000080213
Encompass
The R-PLEX Human CD81 EV Assay Kit from Meso Scale Discovery is a multiplex immunoassay kit for the detection and quantification of CD81-positive extracellular vesicles in human samples The kit contains a 96-well plate pre-coated with anti-CD81 capture antibody detection antibody standards buffers and substrate The assay utilizes electrochemiluminescence technology for high sensitivity and broad dynamic range It can be used for biomarker discovery and characterization of extracellular vesicles from cell culture supernatant plasma serum urine cerebrospinal fluid and other biofluids The kit provides components for 48 tests in duplicate
Catalog No. 50-000-80213 Supplier Meso Scale Discovery Supplier No. K1515NR2
Add to Cart
Add to Cart
The R-PLEX Human CD81 EV Assay Kit from Meso Scale Discovery is a multiplex immunoassay kit for the detection and quantification of CD81-positive extracellular vesicles in human samples The kit contains a 96-well plate pre-coated with anti-CD81 capture antibody detection antibody standards buffers and substrate The assay utilizes electrochemiluminescence technology for high sensitivity and broad dynamic range It can be used for biomarker discovery and characterization of extracellular vesicles from cell culture supernatant plasma serum urine cerebrospinal fluid and other biofluids The kit provides components for 48 tests in duplicate