Learn More
Meso Scale Discovery R-PLEX Human CD81 EV ASYQP
SDP
Catalog No. 5000078898
Encompass
The R-Plex Human CD81 EV Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for detecting and quantifying extracellular vesicles (EVs) with CD81 tetraspanin protein It uses electrochemiluminescent detection for rapid high-sensitivity profiling of EVs from cell culture supernatants biofluids or other samples The kit includes a 96-well plate with anti-CD81 capture antibody detection antibody standards buffers and substrate The detection limit is 50 EVs/well aiding in biomarker discovery and biological studies
Catalog No. 50-000-78898 Supplier Meso Scale Discovery Supplier No. K1515NR22
The R-Plex Human CD81 EV Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for detecting and quantifying extracellular vesicles (EVs) with CD81 tetraspanin protein It uses electrochemiluminescent detection for rapid high-sensitivity profiling of EVs from cell culture supernatants biofluids or other samples The kit includes a 96-well plate with anti-CD81 capture antibody detection antibody standards buffers and substrate The detection limit is 50 EVs/well aiding in biomarker discovery and biological studies