Learn More
Meso Scale Discovery R-PLEX Human CD9 EV ASY
SDP
Catalog No. 5000080560
Encompass
The R-PLEX Human CD9 EV Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the detection and quantitation of CD9-positive extracellular vesicles in human biological samples The kit contains a 96-well plate coated with anti-CD9 capture antibody detection antibody conjugated to an electrochemiluminescent label and calibrator to generate a standard curve The assay has a detection range of 50-50 000 EVs/well and takes 3 5 hours to complete It is suitable for quantifying CD9-positive EVs from cell culture supernatants urine plasma serum and other biofluids The kit provides a sensitive and high-throughput method to enumerate EVs for biomarker research
Catalog No. 50-000-80560 Supplier Meso Scale Discovery Supplier No. K1515MR2
The R-PLEX Human CD9 EV Assay Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the detection and quantitation of CD9-positive extracellular vesicles in human biological samples The kit contains a 96-well plate coated with anti-CD9 capture antibody detection antibody conjugated to an electrochemiluminescent label and calibrator to generate a standard curve The assay has a detection range of 50-50 000 EVs/well and takes 3 5 hours to complete It is suitable for quantifying CD9-positive EVs from cell culture supernatants urine plasma serum and other biofluids The kit provides a sensitive and high-throughput method to enumerate EVs for biomarker research