Learn More
Meso Scale Discovery R-PLEX Human CD9 EV ASYQP
SDP
Catalog No. 5000078413
Encompass
The R-PLEX Human CD9 EV Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures CD9 a tetraspanin protein commonly enriched on the surface of extracellular vesicles (EVs) including exosomes The assay utilizes electrochemiluminescence (ECL) detection and provides a tool to enumerate EVs and monitor EV production The kit includes a 96-well plate pre-coated with anti-CD9 capture antibody detection antibody assay diluent wash buffer read buffer and calibrator The assay has a typical lower limit of detection of 50 EVs/ L and a working range up to 50 000 EVs/ L
Catalog No. 50-000-78413 Supplier Meso Scale Discovery Supplier No. K1515MR22
Add to Cart
Add to Cart
The R-PLEX Human CD9 EV Assay Kit from Meso Scale Discovery is a multiplex immunoassay that quantitatively measures CD9 a tetraspanin protein commonly enriched on the surface of extracellular vesicles (EVs) including exosomes The assay utilizes electrochemiluminescence (ECL) detection and provides a tool to enumerate EVs and monitor EV production The kit includes a 96-well plate pre-coated with anti-CD9 capture antibody detection antibody assay diluent wash buffer read buffer and calibrator The assay has a typical lower limit of detection of 50 EVs/ L and a working range up to 50 000 EVs/ L