Learn More
Meso Scale Discovery R-PLEX Human CEACAM-1 ASY
SDP
Catalog No. 5000078676
Encompass
The Meso Scale Discovery R-PLEX Human CEACAM-1 Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of CEACAM-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and has a testing time of 3 5 hours The lower limit of detection is 1 15 pg/mL It is available as a kit including a 96-well plate coated with capture antibodies detection antibodies calibrators and controls This assay can aid research on cancer inflammation liver disease and diabetes
Catalog No. 50-000-78676 Supplier Meso Scale Discovery Supplier No. K151L8R2
The Meso Scale Discovery R-PLEX Human CEACAM-1 Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of CEACAM-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and has a testing time of 3 5 hours The lower limit of detection is 1 15 pg/mL It is available as a kit including a 96-well plate coated with capture antibodies detection antibodies calibrators and controls This assay can aid research on cancer inflammation liver disease and diabetes