Learn More
Meso Scale Discovery R-PLEX Human CEACAM-1 ASYQP
SDP
Catalog No. 5000079159
Encompass
The R-PLEX Human CEACAM-1 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the simultaneous quantitative measurement of CEACAM-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators diluent and wash buffer It can accurately quantify CEACAM-1 levels between 0 61 pg/mL to 2500 pg/mL
Catalog No. 50-000-79159 Supplier Meso Scale Discovery Supplier No. K151L8R22
The R-PLEX Human CEACAM-1 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the simultaneous quantitative measurement of CEACAM-1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound calibrators diluent and wash buffer It can accurately quantify CEACAM-1 levels between 0 61 pg/mL to 2500 pg/mL