Learn More
Meso Scale Discovery R-PLEX Human CEACAM-5 ASYQP
SDP
Catalog No. 5000078583
Encompass
The R-PLEX Human CEA Assay is a multiplex immunoassay panel that measures Carcinoembryonic Antigen (CEA) in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescence (ECL) detection technology and is compatible with the MSD QuickPlex SQ 120 instrument This assay provides quantitative determination of CEA a tumor marker that can be elevated in gastrointestinal lung breast and other cancers With high sensitivity and broad dynamic range it allows accurate CEA quantification for cancer screening and monitoring The R-PLEX panel is rigorously tested to ensure optimal performance and reliability
Catalog No. 50-000-78583 Supplier Meso Scale Discovery Supplier No. K151QER22
The R-PLEX Human CEA Assay is a multiplex immunoassay panel that measures Carcinoembryonic Antigen (CEA) in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescence (ECL) detection technology and is compatible with the MSD QuickPlex SQ 120 instrument This assay provides quantitative determination of CEA a tumor marker that can be elevated in gastrointestinal lung breast and other cancers With high sensitivity and broad dynamic range it allows accurate CEA quantification for cancer screening and monitoring The R-PLEX panel is rigorously tested to ensure optimal performance and reliability