Learn More
Meso Scale Discovery R-PLEX Human CEACAM-7/CGM2 ASY
SDP
Catalog No. 5000079296
Encompass
The Meso Scale Discovery R-PLEX Human CEACAM-7 Assay is a multiplex immunoassay that measures CEACAM-7 in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity a wide dynamic range and low sample volume This assay quantifies CEACAM-7 a molecule expressed in gastrointestinal tissue and overexpressed in colorectal cancers aiding in the detection and monitoring of colorectal cancer The kit includes a 96-well plate capture and detection antibodies calibrators and controls
Catalog No. 50-000-79296 Supplier Meso Scale Discovery Supplier No. K151M8R2
The Meso Scale Discovery R-PLEX Human CEACAM-7 Assay is a multiplex immunoassay that measures CEACAM-7 in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity a wide dynamic range and low sample volume This assay quantifies CEACAM-7 a molecule expressed in gastrointestinal tissue and overexpressed in colorectal cancers aiding in the detection and monitoring of colorectal cancer The kit includes a 96-well plate capture and detection antibodies calibrators and controls