Learn More
Meso Scale Discovery R-PLEX Human Chemerin ASYQP
SDP
Catalog No. 5000079456
Encompass
The Meso Scale Discovery R-PLEX Human Chemerin Assay is a multiplex immunoassay that measures chemerin levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less of sample and can be run on the MSD QuickPlex SQ 120 MESO QuickPlex SQ 120 or standard SECTOR Instruments The assay measures chemerin an adipokine involved in immune response metabolism and inflammation This assay allows for the quantitative measurement of chemerin with accuracy and reproducibility
Catalog No. 50-000-79456 Supplier Meso Scale Discovery Supplier No. K151ACBR22
The Meso Scale Discovery R-PLEX Human Chemerin Assay is a multiplex immunoassay that measures chemerin levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less of sample and can be run on the MSD QuickPlex SQ 120 MESO QuickPlex SQ 120 or standard SECTOR Instruments The assay measures chemerin an adipokine involved in immune response metabolism and inflammation This assay allows for the quantitative measurement of chemerin with accuracy and reproducibility