Learn More
Meso Scale Discovery R-PLEX Human cIAP1 ASY
SDP
Catalog No. 5000078912
Encompass
The R-Plex Human CIAP1 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures the levels of Cellular Inhibitor of Apoptosis Protein 1 (CIAP1) in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can monitor CIAP1 levels across four orders of magnitude from sub-picogram to nanogram levels per mL This quantitative immunoassay allows the study of CIAP1 levels in biological samples for life science research
Catalog No. 50-000-78912 Supplier Meso Scale Discovery Supplier No. K1517VR2
The R-Plex Human CIAP1 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures the levels of Cellular Inhibitor of Apoptosis Protein 1 (CIAP1) in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can monitor CIAP1 levels across four orders of magnitude from sub-picogram to nanogram levels per mL This quantitative immunoassay allows the study of CIAP1 levels in biological samples for life science research